Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Benzofluorene 97.0+%, TCI America™
SDP

Catalog No. B00591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00591G 1 g
B0059100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B00591G Supplier TCI America Supplier No. B00591G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 243-17-4
Molecular Formula C17H12
Molecular Weight (g/mol) 216.28
MDL Number MFCD00003595
InChI Key HAPOJKSPCGLOOD-UHFFFAOYSA-N
Synonym 2,3-benzofluorene, 11h-benzo b fluorene, benzo b fluorene, unii-gmo75ruy0l, 2,3-benzfluorene, gmo75ruy0l, 11h-benzo b fluorene #
PubChem CID 9201
ChEBI CHEBI:34566
IUPAC Name 11H-benzo[b]fluorene
SMILES C1C2=CC=CC=C2C2=C1C=C1C=CC=CC1=C2

Specifications

Melting Point 213°C
Color White-Yellow
Boiling Point 402°C
Quantity 1 g
Formula Weight 216.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Benzofluorene
RTECSNumber DF6383500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.