Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(4-ethylphenyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™
SDP

Catalog No. B1053100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1053100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1053100MG Supplier TCI America Supplier No. B1053100MG

Chemical Identifiers

CAS 104497-78-1
Molecular Formula C23H23ClN4
Molecular Weight (g/mol) 390.92
MDL Number MFCD00060006
InChI Key PQLZEEMSIAJGGW-UHFFFAOYSA-M

Specifications

Quantity 100 mg
Formula Weight 390.92
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-ethylphenyl)-5-phenyltetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.