Learn More
Medchemexpress LLC 2,3-bis(4-hydroxyphenyl)propionitrile | 1428-67-7 | MFCD01695412 | >98.0% | 239.27 g/mol | C15H13NO2 | 500MG
SDP
Catalog No. 5000237034
Encompass_Preferred
DPN (diarylpropionitrile) is a synthetic, non-steroidal estrogen receptor β (ERβ) selective agonist used as a research reagent. It has a reported EC50 of 0.85 nM for ERβ and is used in studies of ERβ pharmacology and neuroprotection. CAS 1428-67-7; formula C15H13NO2; molecular weight 239.27 g/mol.
  • Selective ERβ agonist with low-nanomolar potency.
  • Reported EC50 of 0.85 nM for ERβ.
  • Suitable for in vitro pharmacology and neuroprotection studies.
  • High purity (>98.0% reported by suppliers).
Catalog No. 50-002-37034 Supplier Medchemexpress LLC Supplier No. HY12452500MG
DPN (diarylpropionitrile) is a synthetic, non-steroidal estrogen receptor β (ERβ) selective agonist used as a research reagent. It has a reported EC50 of 0.85 nM for ERβ and is used in studies of ERβ pharmacology and neuroprotection. CAS 1428-67-7; formula C15H13NO2; molecular weight 239.27 g/mol.
  • Selective ERβ agonist with low-nanomolar potency.
  • Reported EC50 of 0.85 nM for ERβ.
  • Suitable for in vitro pharmacology and neuroprotection studies.
  • High purity (>98.0% reported by suppliers).