Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(4-methylphenyl)-5-(4-cyanophenyl)tetrazolium Chloride 95.0+%, TCI America™
SDP

Catalog No. B1312100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1312100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B1312100MG Supplier TCI America Supplier No. B1312100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 376591-03-6
Molecular Formula C22H18ClN5
Molecular Weight (g/mol) 387.871
MDL Number MFCD00060130
InChI Key MEDRXJSBVPMSHT-UHFFFAOYSA-M
PubChem CID 44630131
IUPAC Name 4-[2,3-bis(4-methylphenyl)tetrazol-2-ium-5-yl]benzonitrile;chloride
SMILES CC1=CC=C(C=C1)N2N=C(N=[N+]2C3=CC=C(C=C3)C)C4=CC=C(C=C4)C#N.[Cl-]

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 387.87
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-methylphenyl)-5-(4-cyanophenyl)tetrazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.