Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(bromomethyl)quinoxaline, 97%, Thermo Scientific™

Catalog No. AAH5864904
Change view
Click to view available options
Quantity:
2 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5864904 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5864904 Supplier Thermo Scientific Chemicals Supplier No. H5864904

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3138-86-1
Molecular Formula C10H8Br2N2
Molecular Weight (g/mol) 315.996
MDL Number MFCD00006729
InChI Key LHKFFORGJVELPC-UHFFFAOYSA-N
Synonym 2,3-bis bromomethyl quinoxaline, quinoxaline, 2,3-bis bromomethyl, 2,3-bis bromomethyl-1,4-benzodiazine, benzoic acid,3-fluoro-5-nitro, 2,4-benzodiazine, 2-hydroxybenzthiazole, acmc-1cq4g, 5-23-07-00271 beilstein handbook reference, ksc522c3j, quinoxaline,3-bis bromomethyl
PubChem CID 18449
IUPAC Name 2,3-bis(bromomethyl)quinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)CBr)CBr

Specifications

Melting Point 152°C to 156°C
Quantity 2 g
UN Number UN3261
Formula Weight 315.99
Percent Purity 97%
Chemical Name or Material 2,3-Bis(bromomethyl)quinoxaline
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 221-538-4
RTECSNumber VD1420000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.