Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(bromomethyl)quinoxaline, 98%, Thermo Scientific™

Catalog No. AC106310100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC106310100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC106310100 Supplier Thermo Scientific Chemicals Supplier No. 106310100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3138-86-1
Molecular Formula C10H8Br2N2
Molecular Weight (g/mol) 315.996
InChI Key LHKFFORGJVELPC-UHFFFAOYSA-N
Synonym 2,3-bis bromomethyl quinoxaline, quinoxaline, 2,3-bis bromomethyl, 2,3-bis bromomethyl-1,4-benzodiazine, benzoic acid,3-fluoro-5-nitro, 2,4-benzodiazine, 2-hydroxybenzthiazole, acmc-1cq4g, 5-23-07-00271 beilstein handbook reference, ksc522c3j, quinoxaline,3-bis bromomethyl
PubChem CID 18449
IUPAC Name 2,3-bis(bromomethyl)quinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)CBr)CBr

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 150°C to 154°C
Color Beige-Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 10 g
Beilstein 23, II, 197
Formula Weight 315.98
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Bis(bromomethyl)quinoxaline, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.