Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(hydroxyamino)-2,3-dimethylbutane Sulfate (Pract.), 85%, Thermo Scientific™

Catalog No. AC402520250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC402520250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402520250 Supplier Thermo Scientific Chemicals Supplier No. 402520250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14538-51-3
Molecular Formula C6H16N2O2.H2SO4
Synonym N,N'-Dihydroxy-2,3-dimethyl-2,3-butanediamine sulfate

Specifications

CAS Min % 80.0
CAS Max % 100.0
Melting Point 170°C to 175°C
Color White
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 246.29
Percent Purity ≥80%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 3-Bis(hydroxyamino)-2, 3-dimethylbutane sulfate, pract., 85%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.