Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(hydroxymethyl)naphthalene 98.0+%, TCI America™
SDP

Catalog No. B15891G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15891G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B15891G Supplier TCI America Supplier No. B15891G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 31554-15-1
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD00191314
InChI Key DTYJRRQQOLBRGH-UHFFFAOYSA-N
PubChem CID 598417
IUPAC Name [3-(hydroxymethyl)naphthalen-2-yl]methanol
SMILES C1=CC=C2C=C(C(=CC2=C1)CO)CO

Specifications

Melting Point 157°C
Color White-Yellow
Quantity 1 g
Formula Weight 188.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(hydroxymethyl)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.