Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(trifluoromethyl)pyridine 95.0+%, TCI America™
SDP

Catalog No. B23941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23941G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23941G Supplier TCI America Supplier No. B23941G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1644-68-4
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD03093763
InChI Key RRNXYHYDSDAOFW-UHFFFAOYSA-N
Synonym 2,3-bis trifluoromethyl pyridine, pyridine, 2,3-bis trifluoromethyl, pubchem7694, acmc-209dra, bis trifluoromethyl pyridine, 2,3-bis trifluoromethyl pyridine #, pyridine,2,3-bis trifluoromethyl
PubChem CID 589865
IUPAC Name 2,3-bis(trifluoromethyl)pyridine
SMILES C1=CC(=C(N=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 2°C
Color Yellow
Boiling Point 158°C
Quantity 1 g
Formula Weight 215.10
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Bis(trifluoromethyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.