Learn More
Medchemexpress LLC 2-[(3-bromo-5-tert-butyl-4-hydroxyphenyl)methylidene]propanedinitrile | 65678-07-1 | MFCD02179365 | >98.0% | 305.17 g/mol | C14H13BrN2O | 100 MG
SDP
Catalog No. 5000247108
Encompass_Preferred
AG1024 (Tyrphostin AG 1024) is a small-molecule, reversible inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) used in cell signaling and cancer research. It inhibits IGF-1R autophosphorylation and can modulate insulin receptor activity; the material is supplied as a powder for laboratory use.
  • Reversible inhibitor of IGF-1R suitable for mechanistic studies.
  • Inhibits IGF-1R autophosphorylation and modulates insulin receptor signaling.
  • High purity suitable for cell-based and biochemical assays.
  • Powder form with good solubility in DMSO for solution preparation.
  • Provided in research-scale quantities for laboratory testing.
Catalog No. 50-002-47108 Supplier Medchemexpress LLC Supplier No. HY10253100MG
AG1024 (Tyrphostin AG 1024) is a small-molecule, reversible inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) used in cell signaling and cancer research. It inhibits IGF-1R autophosphorylation and can modulate insulin receptor activity; the material is supplied as a powder for laboratory use.
  • Reversible inhibitor of IGF-1R suitable for mechanistic studies.
  • Inhibits IGF-1R autophosphorylation and modulates insulin receptor signaling.
  • High purity suitable for cell-based and biochemical assays.
  • Powder form with good solubility in DMSO for solution preparation.
  • Provided in research-scale quantities for laboratory testing.