Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine 97.0+%, TCI America™
SDP

Catalog No. B48941G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48941G 1 g
B48945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48941G Supplier TCI America Supplier No. B48941G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 864377-31-1
Molecular Formula C21H14BrN3
Molecular Weight (g/mol) 388.27
MDL Number MFCD22200060
InChI Key HNZUKQQNZRMNGS-UHFFFAOYSA-N
PubChem CID 58943302
IUPAC Name 2-(3-bromophenyl)-4,6-diphenyl-1,3,5-triazine
SMILES BrC1=CC=CC(=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 176°C
Color White
Quantity 1 g
Formula Weight 388.27
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Bromophenyl)-4,6-diphenyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.