Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Bromophenyl)pyridine, 97%, Thermo Scientific™

Catalog No. AC457560050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC457560050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC457560050 Supplier Thermo Scientific Chemicals Supplier No. 457560050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4373-60-8
Molecular Formula C11H8BrN
Molecular Weight (g/mol) 234.1
InChI Key WLPFTJXVEBANAM-UHFFFAOYSA-N
Synonym 2-3-bromophenyl pyridine, pyridine, 2-3-bromophenyl, 2-3-bromo-phenyl-pyridine, 2-3'-bromophenyl pyridine, 3-2-pyridinyl phenyl bromide, 3,5-pyrid-2-yl bromobenzene
PubChem CID 11115506
IUPAC Name 2-(3-bromophenyl)pyridine
SMILES C1=CC=NC(=C1)C2=CC(=CC=C2)Br

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 234.1
Percent Purity 97%
Chemical Name or Material 2-(3-Bromophenyl)pyridine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.