Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Bromophenyl)triphenylene 98.0+%, TCI America™
SDP

Catalog No. B47461G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47461G 1 g
B4746200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47461G Supplier TCI America Supplier No. B47461G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1313514-53-2
Molecular Formula C24H15Br
Molecular Weight (g/mol) 383.288
InChI Key KWXFBIBEVROWEF-UHFFFAOYSA-N
PubChem CID 58472032
IUPAC Name 2-(3-bromophenyl)triphenylene
SMILES C1=CC=C2C(=C1)C3=C(C=C(C=C3)C4=CC(=CC=C4)Br)C5=CC=CC=C25

Specifications

Melting Point 149°C
Color White-Yellow
Quantity 1 g
Formula Weight 383.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Bromophenyl)triphenylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.