Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-(3-But-3-Ynyl-3H-Diazirin-3-Yl)-Ethanol

Catalog No. 90086350MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-086-350MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-086-350MG Supplier Sigma Aldrich Supplier No. 90086350MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

2-(3-But-3-ynyl-3H-diazirin-3-yl)-ethanol is used for chemical probe synthesis, this trifunctional building block contains a light-activated diazirine, alkyne tag, and hydroxyl synthetic handle. When appended to a ligand or pharmacophore through its hydroxyl linker, this building block allows for UV light-induced covalent modification of a biological target with potential for downstream applications via the alkyne tag. Use alone or in parallel with other multi-functional building blocks to discover the optimal probe for your chemical biology experiments.Technology Spotlight: Trifunctional Probe Building BlocksFluorescent Probes for Single-Step Detection and Proteomic Profiling of Histone DeacetylasesIn Situ Proteome Profiling and Bioimaging Applications of Small-Molecule Affinity-Based Probes Derived From DOT1L InhibitorsReversible inhibitors arrest ClpP in a defined conformational state that can be revoked by ClpX associationDesign and Synthesis of Minimalist Terminal Alkyne-Containing Diazirine Photo-Crosslinkers and Their Incorporation into Kinase Inhibitors for Cell- and Tissue-Based Proteome Profiling

Specifications

CAS 653600-56-7
Molecular Formula C16H22N4O10
Linear Formula C16H22N4O10
Quantity 50 mg
Synonym Ac4GalNAz
Molecular Weight (g/mol) 430.37 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.