Learn More
Medchemexpress LLC 2,3-Butanediol | 513-85-9 | 100.0% | 10 MM 1 ML
SDP
Catalog No. 5000339240
Encompass_Preferred
2,3-Butanediol is a microbially derived compound and a discriminating urinary biomarker for Fmo5-/- mice. It prevents age-related increases in plasma cholesterol concentration and has potential applications in various industries, including printing inks, perfumes, fumigants, moistening and softening agents, and plasticizers, as well as serving as a pharmaceutical carrier. This compound features two stereo centers, leading to three distinct stereo isomers: (2R,3R)-, meso- and (2S,3S)-forms.
  • Functions as a discriminating urinary biomarker for Fmo5-/- mice.
  • Helps prevent age-related increases in plasma cholesterol.
  • Applicable in manufacturing printing inks, perfumes, and fumigants.
  • Useful as a moistening and softening agent, plasticizer, or pharmaceutical carrier.
  • Composed of three stereo isomers due to two stereo centers.
  • Demonstrated to lower plasma cholesterol and epididymal fat deposition in studies.
Catalog No. 50-003-39240 Supplier Medchemexpress LLC Supplier No. HY12838710MM/1ML
2,3-Butanediol is a microbially derived compound and a discriminating urinary biomarker for Fmo5-/- mice. It prevents age-related increases in plasma cholesterol concentration and has potential applications in various industries, including printing inks, perfumes, fumigants, moistening and softening agents, and plasticizers, as well as serving as a pharmaceutical carrier. This compound features two stereo centers, leading to three distinct stereo isomers: (2R,3R)-, meso- and (2S,3S)-forms.
  • Functions as a discriminating urinary biomarker for Fmo5-/- mice.
  • Helps prevent age-related increases in plasma cholesterol.
  • Applicable in manufacturing printing inks, perfumes, and fumigants.
  • Useful as a moistening and softening agent, plasticizer, or pharmaceutical carrier.
  • Composed of three stereo isomers due to two stereo centers.
  • Demonstrated to lower plasma cholesterol and epididymal fat deposition in studies.