Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butanediol, ca. 85% meso, Remainder DL (Tech.), Thermo Scientific™

Catalog No. AC332022500
Change view
Click to view available options
Quantity:
250 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC332022500 250 mL
AC332020010 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC332022500 Supplier Thermo Scientific Chemicals Supplier No. 332022500

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-85-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
InChI Key OWBTYPJTUOEWEK-UHFFFAOYSA-N
Synonym 2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
PubChem CID 262
ChEBI CHEBI:62064
IUPAC Name butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

Color Colorless to Yellow
Infrared Spectrum Authentic
Refractive Index 1.4360 to 1.4400 (20°C, 589nm)
Linear Formula CH3CH(OH)CH(OH)CH3
Quantity 250 mL
Beilstein 01, 479
Merck Index 15, 1571
Formula Weight 90.12
Percent Purity ≥80%
Grade Technical
Physical Form Viscous Liquid
Chemical Name or Material 2, 3-Butanediol, tech.
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 208-173-6
RTECSNumber EK0532000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.