Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butanediol, Mixture of racemic and meso forms, ≥97.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014403 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4403 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4403 Supplier MilliporeSigma Supelco Supplier No. 42038250MG
Add to Cart
Add to Cart

2,3-Butanediol is a chiral compound which is extensively utilized in chemical feedstocks and liquid fuels.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 513-85-9
Density 1.002 g/mL (at 20°C)
Flash Point 85°C
Physical Form Neat
Linear Formula CH3 CH(OH)CH(OH)CH3
MDL Number MFCD00004523
Melting Point 25°C
Boiling Point 183°C to 184°C
Synonym 2,3-Butylene glycol
Type Analytical Standard
Quantity 250 mg
Formula Weight 90.12
Percent Purity ≥97.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.