Learn More
Medchemexpress LLC 2,3-Butanediol (Standard) | 513-85-9 | 99.8% | 50 MG
SDP
Catalog No. 5000339071
Encompass_Preferred
2,3-Butanediol (Standard) is the analytical standard of 2,3-Butanediol. This product is intended for research and analytical applications. 2,3-Butanediol is a microbially derived compound that is a discriminating urinary biomarker of Fmo5-/- mice. It also prevents age-related increases in the plasma concentration of cholesterol.
  • Potential applications in printing inks manufacturing
  • Potential applications in perfumes manufacturing
  • Potential applications in fumigants manufacturing
  • Potential applications as moistening and softening agents
  • Potential applications as plasticizers
  • Potential applications as a carrier for pharmaceuticals
Catalog No. 50-003-39071 Supplier Medchemexpress LLC Supplier No. HY128387R50MG
2,3-Butanediol (Standard) is the analytical standard of 2,3-Butanediol. This product is intended for research and analytical applications. 2,3-Butanediol is a microbially derived compound that is a discriminating urinary biomarker of Fmo5-/- mice. It also prevents age-related increases in the plasma concentration of cholesterol.
  • Potential applications in printing inks manufacturing
  • Potential applications in perfumes manufacturing
  • Potential applications in fumigants manufacturing
  • Potential applications as moistening and softening agents
  • Potential applications as plasticizers
  • Potential applications as a carrier for pharmaceuticals