Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2,3-Butanedione

Catalog No. B853075ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B853075ML 5 mL
B85307500ML 500 mL
B85307100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B853075ML Supplier Sigma Aldrich Supplier No. B853075ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Inactivates aminopeptidase-n
  • Cyclocondensation with amines has been used to form triazine and pteridine ring systems
  • Also used as a precursor to α-diones

Specifications

CAS 431-03-8
Density 0.981 g/mL (literature)
Boiling Point 88°C (lit.)
Molecular Formula C4H6O2
Refractive Index n20/D 1.394 (literature)
Linear Formula CH3COCOCH3
MDL Number MFCD00008756
Quantity 5 mL
Synonym Biacetyl; Diacetyl
Molecular Weight (g/mol) 86.09
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.