Learn More
Selleck Chemical LLC 2.3-Butanedione-2-monoxime S3699-1g
SDP
Catalog No. 503109737
Encompass_Preferred
2 3-Butanedione monoxime (BDM Diacetyl monoxime Diacetylmonoxime) is the well-characterized low-affinity non-competitive inhibitor of skeletal muscle myosin-II and inhibits skeletal and cardiac muscle contraction
Catalog No. 50-310-9737 Supplier Selleck Chemical LLC Supplier No. S36991G
2 3-Butanedione monoxime (BDM Diacetyl monoxime Diacetylmonoxime) is the well-characterized low-affinity non-competitive inhibitor of skeletal muscle myosin-II and inhibits skeletal and cardiac muscle contraction