Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butanedione, Analytical Standard, ≥99.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019163 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9163 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9163 Supplier MilliporeSigma Supelco Supplier No. 110385MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 431-03-8
Vapor Pressure 52.2 mmHg (20°C)
Vapor Density 3 (vs. air)
Density 0.981 g/mL
Physical Form Neat
Linear Formula CH3COCOCH3
MDL Number MFCD00008756
Boiling Point 88°C
Synonym Biacetyl; Diacetyl
Quantity 5 mL
Formula Weight 86.09
Percent Purity ≥99.0% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.