Learn More
Selleck Chemical LLC 2,3-Butanedione-2-monoxime 25mg 57-71-6 Diacetyl monoxime, Diacetylmonoxime
SDP
Supplier:  Selleck Chemical LLC S369925MG
Encompass_Preferred
2,3-Butanedione monoxime (BDM, Diacetyl monoxime, Diacetylmonoxime) is the well-characterized, low-affinity, non-competitive inhibitor of skeletal muscle myosin-II and inhibits skeletal and cardiac muscle contraction. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8346