Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butanedione monoxime, For Spectrophotometric Det. of Urea, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111014723 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4723 100 g
11-101-4722 25 g
11-101-4721 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-4723 Supplier MilliporeSigma Supelco Supplier No. 31550100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

MDL Number MFCD00002116
Synonym BDM; Biacetyl monoxime; Diacetyl monoxime

Specifications

Melting Point 75°C to 76°C; 75°C to 78°C
CAS 57-71-6
Boiling Point 185°C to 186°C
Linear Formula CH3 C(=NOH)COCH3
Quantity 100 g
Formula Weight 101.10
Percent Purity ≥98.0% (N); ≥99.0%
RTECSNumber EK3150000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.