Learn More
Cayman Chemical 2,3-Butanedione-2-monoxime, Purity: ≥95%, CAS: 57-71-6, 10 g
SDP
Catalog No. 502984084
Encompass_Preferred
A non-selective, reversible myosin ATPase inhibitor with phosphatase-like properties; inhibits L-type Ca2+ channels
Catalog No. 50-298-4084 Supplier Cayman Chemical Supplier No. 2082810 G
A non-selective, reversible myosin ATPase inhibitor with phosphatase-like properties; inhibits L-type Ca2+ channels