Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Butylene Oxide (cis- and trans- mixture), TCI America™
SDP

Catalog No. B072010ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B072010ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B072010ML Supplier TCI America Supplier No. B072010ML

Chemical Identifiers

CAS 3266-23-7
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00066345
InChI Key PQXKWPLDPFFDJP-UHFFFAOYSA-N
Synonym 2,3-Epoxybutane, Pseudobutylene Oxide
PubChem CID 18632
IUPAC Name 2,3-dimethyloxirane
SMILES CC1C(O1)C

Specifications

Color Colorless
UN Number 1993
Quantity 10 mL
Formula Weight 72.11
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Butylene Oxide (cis- and trans- mixture)
EINECSNumber (2)-0229
RTECSNumber EK3855000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.