Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2'3'-c-di-AM(PS)2 (Rp,Rp) enantiomer (ammonium salt) | 98.9% | 10 MG
SDP

Catalog No. 5000339823
Encompass_Preferred

2'3'-c-di-AM(PS)2 (Rp,Rp) enantiomer ammonium salt is the less active enantiomer of 2'3'-c-di-AM(PS)2 (Rp,Rp), an activator of stimulator of interferon genes (STING). This chemical is intended for laboratory research and manufacture of substances.

  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 724.60
  • Formula: C20H30N12O10P2S2
  • Purity: 98.9%
  • Solubility in water: ≥ 100 mg/mL
  • Solubility in DMSO: 50 mg/mL
  • Recommended storage for solid: -20°C, sealed, away from moisture and light.

Catalog No. 50-003-39823 Supplier Medchemexpress LLC Supplier No. HY12885C10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2'3'-c-di-AM(PS)2 (Rp,Rp) enantiomer ammonium salt is the less active enantiomer of 2'3'-c-di-AM(PS)2 (Rp,Rp), an activator of stimulator of interferon genes (STING). This chemical is intended for laboratory research and manufacture of substances.

  • Appearance: Solid
  • Color: White to off-white
  • Molecular weight: 724.60
  • Formula: C20H30N12O10P2S2
  • Purity: 98.9%
  • Solubility in water: ≥ 100 mg/mL
  • Solubility in DMSO: 50 mg/mL
  • Recommended storage for solid: -20°C, sealed, away from moisture and light.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.