Learn More
Medchemexpress LLC 2-[3-carboxy-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic acid | 1239233-87-4 | MFCD29050077 | 98.8% | 335.37 | C16H17NO5S | 5 MG
SDP
Catalog No. 5000263028
Encompass_Preferred
Febuxostat dicarboxylic acid impurity is an analytical reference impurity derived from the xanthine oxidase inhibitor febuxostat, provided for research and quality-control applications. It is intended for use in analytical characterization, method development, and impurity profiling in pharmaceutical analysis.
  • Analytical reference impurity for febuxostat.
  • High purity for reliable analytical results.
  • Suitable for method development and impurity profiling.
  • Provided with supporting analytical documentation such as COA.
  • Supplied in small, research-grade quantities for laboratory use.
Catalog No. 50-002-63028 Supplier Medchemexpress LLC Supplier No. HY1312725MG
Febuxostat dicarboxylic acid impurity is an analytical reference impurity derived from the xanthine oxidase inhibitor febuxostat, provided for research and quality-control applications. It is intended for use in analytical characterization, method development, and impurity profiling in pharmaceutical analysis.
  • Analytical reference impurity for febuxostat.
  • High purity for reliable analytical results.
  • Suitable for method development and impurity profiling.
  • Provided with supporting analytical documentation such as COA.
  • Supplied in small, research-grade quantities for laboratory use.