Learn More
Medchemexpress LLC 2-[3-carboxy-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic acid | 1239233-87-4 | MFCD29050077 | 98.8% | C16H17NO5S | 10MG
SDP
Catalog No. 5000260676
Encompass_Preferred
Febuxostat dicarboxylic acid impurity is an analytical reference material used for impurity profiling and method development related to febuxostat. It is provided for research and analytical applications, with CAS 1239233-87-4, molecular formula C16H17NO5S, molecular weight 335.37 g·mol⁻¹, and a typical assay of 98.8%.
  • For use as an analytical impurity standard in method development and validation.
  • High purity suitable for HPLC and mass spectrometry analysis.
  • Provided in milligram-scale packaging for reference and QC work.
  • Molecular formula C16H17NO5S and CAS 1239233-87-4 for unambiguous identification.
  • Certificate of analysis and batch data available to support traceability.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-60676 Supplier Medchemexpress LLC Supplier No. HY13127210MG
Febuxostat dicarboxylic acid impurity is an analytical reference material used for impurity profiling and method development related to febuxostat. It is provided for research and analytical applications, with CAS 1239233-87-4, molecular formula C16H17NO5S, molecular weight 335.37 g·mol⁻¹, and a typical assay of 98.8%.
  • For use as an analytical impurity standard in method development and validation.
  • High purity suitable for HPLC and mass spectrometry analysis.
  • Provided in milligram-scale packaging for reference and QC work.
  • Molecular formula C16H17NO5S and CAS 1239233-87-4 for unambiguous identification.
  • Certificate of analysis and batch data available to support traceability.
  • Intended for research use only; not for human or clinical use.