Learn More
Invivogen 2'3'-cGAMP, Mammalian (non-canonical) CDN, cyclic [G(2’,5’)pA(3’,5’)p], 1441190-66-4, MFCD31563596, 200 µg
SDP
Catalog No. NC1660053
Encompass
Formula: C20H22N10O13P2 .2Na, Purity: ≥ 95% by LC/MS, Molecular weight: 718.38, 2’3’-cGAMP (cyclic [G(2’,5’)pA(3’,5’)p]) is produced in mammalian cells by cGAS (cGAMP synthase) in response to double-stranded DNA in the cytoplasm. It is also referred to as “noncanonical“ cGAMP due to the presence of the atypical 2’-5’ phosphodiester linkage between the guanosine and the adenosine.
Catalog No. NC1660053 Supplier Invivogen Supplier No. TLRLNACGA2302
Formula: C20H22N10O13P2 .2Na, Purity: ≥ 95% by LC/MS, Molecular weight: 718.38, 2’3’-cGAMP (cyclic [G(2’,5’)pA(3’,5’)p]) is produced in mammalian cells by cGAS (cGAMP synthase) in response to double-stranded DNA in the cytoplasm. It is also referred to as “noncanonical“ cGAMP due to the presence of the atypical 2’-5’ phosphodiester linkage between the guanosine and the adenosine.