Learn More
Invivogen 2'3'-cGAMP Control, 1 mg
SDP
Catalog No. NC1064193
Encompass
2’3’-cGAMP Control, also known as 2’5’-GpAp, is a linear dinucleotide analog after hydrolysis of cyclic guanosine monophosphate- adenosine monophosphate (2’3’-cGAMP) by phosphodiesterases.
Catalog No. NC1064193 Supplier Invivogen Supplier No. TLRLNAGPAP
2’3’-cGAMP Control, also known as 2’5’-GpAp, is a linear dinucleotide analog after hydrolysis of cyclic guanosine monophosphate- adenosine monophosphate (2’3’-cGAMP) by phosphodiesterases.