Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 2',3'-cGAMP sodium | 2734858-36-5 | 99.9% | 718.37 | C20H22N10Na2O13P2 | 10 MG
SDP

Catalog No. 5000211635
Encompass_Preferred

2',3'-cGAMP sodium is the sodium salt of the endogenous cyclic dinucleotide 2',3'-cyclic GMP-AMP (cGAMP). It activates the STING pathway and is used as a research reagent to study innate immune signaling and interferon responses.

  • Activates STING to induce interferon signaling.
  • High reported purity (~99.9%).
  • Molecular weight 718.37 g/mol.
  • Soluble in water (≈50 mg/mL) and PBS; limited solubility in DMSO.
  • Recommended storage at -20°C; in solution store at -80°C for long-term stability.
  • Available in small research pack sizes such as 10 MG vials.

Catalog No. 50-002-11635 Supplier Medchemexpress LLC Supplier No. HY100564A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2',3'-cGAMP sodium is the sodium salt of the endogenous cyclic dinucleotide 2',3'-cyclic GMP-AMP (cGAMP). It activates the STING pathway and is used as a research reagent to study innate immune signaling and interferon responses.

  • Activates STING to induce interferon signaling.
  • High reported purity (~99.9%).
  • Molecular weight 718.37 g/mol.
  • Soluble in water (≈50 mg/mL) and PBS; limited solubility in DMSO.
  • Recommended storage at -20°C; in solution store at -80°C for long-term stability.
  • Available in small research pack sizes such as 10 MG vials.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.