Learn More
Medchemexpress LLC HY-100564A 5mg , 2',3'-cGAMP (sodium) CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100564A5MG
Encompass_Preferred
HY-100564A 5mg 2',3'-cGAMP (sodium) CAS: 2',3'-cGAMP sodium (2'-3'-cyclic GMP-AMP sodium) is a endogenous cGAMP in mammalian cells. 2',3'-cGAMP sodium binds to STING with a high affinity and is a potent inducer of interferon-β (IFNβ). 2',3'-cGAMP sodium is produced in mammalian cells in response to DNA in the cytoplasm. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9591