Learn More
Invivogen Mammalian (non-canonical) CDN, cyclic (G(2’,5’)pA(3’,5’)p), 5mg
SDP
Catalog No. NC1142808
Encompass
2’3’-cGAMP (cyclic (G(2’,5’)pA(3’,5’)p)) is produced in mammalian cells by cGAS (cGAMP synthase) in response to double-stranded DNA in the cytoplasm.
Catalog No. NC1142808 Supplier Invivogen Supplier No. TLRLNACGA235
2’3’-cGAMP (cyclic (G(2’,5’)pA(3’,5’)p)) is produced in mammalian cells by cGAS (cGAMP synthase) in response to double-stranded DNA in the cytoplasm.