Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3'-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine 98.0+%, TCI America™
SDP

Catalog No. C3162200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3162200MG 200 mg
C31621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3162200MG Supplier TCI America Supplier No. C3162200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1443049-83-9
Molecular Formula C27H18ClN3
Molecular Weight (g/mol) 419.912
InChI Key MMAIMCOMSPMTKJ-UHFFFAOYSA-N
PubChem CID 90177101
IUPAC Name 2-[3-(3-chlorophenyl)phenyl]-4,6-diphenyl-1,3,5-triazine
SMILES C1=CC=C(C=C1)C2=NC(=NC(=N2)C3=CC=CC(=C3)C4=CC(=CC=C4)Cl)C5=CC=CC=C5

Specifications

Melting Point 194°C
Color White-Yellow
Quantity 200 mg
Formula Weight 419.91
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3′-Chlorobiphenyl-3-yl)-4,6-diphenyl-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.