Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Chlorophenyl)benzimidazole, 95%, Thermo Scientific™

Catalog No. AAH64245MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH64245MD 250 mg
AAH6424503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH64245MD Supplier Thermo Scientific Chemicals Supplier No. H64245MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22868-35-5
Molecular Formula C13H9ClN2
Molecular Weight (g/mol) 228.68
MDL Number MFCD01438282
InChI Key BGYHTIIVIGGCIF-UHFFFAOYSA-N
PubChem CID 767074
SMILES ClC1=CC=CC(=C1)C1=NC2=CC=CC=C2N1

Specifications

Quantity 250 mg
Synonym 2-3-chlorophenyl-1h-benzimidazole, 2-3-chlorophenyl benzimidazole, 2-3-chlorophenyl-1h-1,3-benzodiazole, 2-3-chlorophenyl-1h-benzo d imidazole, 2-3-chloro-phenyl-1h-benzoimidazole, 1h-benzimidazole, 2-3-chlorophenyl, cbmicro_048649, 1h-benzimidazole,2-3-chlorophenyl
IUPAC Name 2-(3-chlorophenyl)-1H-benzimidazole
Formula Weight 228.68
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 2-(3-Chlorophenyl)benzimidazole
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.