Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 2'3'- Cyclic GAMP ELISA Kit

Catalog No. EIAGAMP
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EIAGAMP 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EIAGAMP Supplier Invitrogen™ Supplier No. EIAGAMP
Add to Cart
Add to Cart

ELISA

The 2',3'-Cyclic GAMP Competitive ELISA Kit quantitates 2',3'-cyclic GAMP in lysed cells and tissue, tissue culture media, and EDTA plasma. Principle of the method: The 2',3'-Cyclic GAMP Competitive ELISA research-use-only kit is designed to quantitatively measure 2',3'-cyclic GAMP independent of species. A 2',3'-cyclic GAMP standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit 2',3'-cyclic GAMP antibodies and 2',3'-cyclic GAMP-peroxidase conjugate is added to the wells. The 2',3'-cyclic GAMP-peroxidase conjugate and any 2',3'-cyclic GAMP in the sample will compete to bind to the anti-rabbit 2',3'-cyclic GAMP antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound 2',3'-cyclic GAMP-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of 2',3'-cyclic GAMP present in the sample. Rigorous validation: Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Cyclic GMP-AMP (cGAMP) synthase (cGAS) is a nucleotidyltransferase located in the cytosol that acts as a cytosolic DNA sensor to detect foreign DNA from microbial pathogens as part of the innate immune response. Upon binding to cytosolic DNA, cGAS produces the cyclic dinucleotide second messenger cGAMP, which activates stimulator of interferon genes (STING), leading to activation of the type I interferon (IFN) pathway. In vitro, fibroblasts, macrophages, and dendritic cells isolated from cGAS knockout (cGAS-/-) mice do not produce type I IFNs following DNA transfection or DNA virus infection. Similarly, cells containing a frame-shift mutation in the cGAS locus fail to mount an immune response to HIV and other retroviruses. In vivo, cGAS-/- mice infected with herpes simplex virus 1 (HSV-1) have lower levels of IFN-α and IFN-β, shorter survival times, and higher post-mortem levels of HSV-1 in the brain.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 2'3 '-Cyclic GMP-AMP; cGAMP; Guanosine-Adenosine 2 ',3 '-cyclic monophosphate
Assay Range 0.08-20 pmol/mL
Assay Sensitivity 0.048 pmol/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Cell Lysates, Plasma, Supernatant, Tissue Homogenates
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 2'3'-Cyclic GMP-AMP; cGAMP; Guanosine-Adenosine 2',3'-cyclic monophosphate2'3'-Cyclic GMP-AMP; cGAMP; Guanosine-Adenosine 2',3'-cyclic monophosphate;
Interassay CV 8%
Intraassay CV 6%
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate, 50 μL; Tissue Homogenate, 50 μL; Supernatant, 50 μL; Plasma, 10 μL
Shipping Condition Wet or Dry Ice
Storage Requirements 2-8°C
Target cGAMP
Test Time 45 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.