Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Cyclopentenopyridine, 99+%, Thermo Scientific™

Catalog No. AC188881000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC188881000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC188881000 Supplier Thermo Scientific Chemicals Supplier No. 188881000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 533-37-9
Molecular Formula C8H9N
Molecular Weight (g/mol) 119.167
InChI Key KRNSYSYRLQDHDK-UHFFFAOYSA-N
Synonym 2,3-cyclopentenopyridine, 6,7-dihydro-5h-cyclopenta b pyridine, pyrindan, 2,3-cyclopenteno pyridine, 5h-1-pyrindine, 6,7-dihydro, 5h,6h,7h-cyclopenta b pyridine, 5h-cyclopenta b pyridine, 6,7-dihydro, pubchem15900, 2,3-cyclopentapyridine
PubChem CID 68292
IUPAC Name 6,7-dihydro-5H-cyclopenta[b]pyridine
SMILES C1CC2=C(C1)N=CC=C2

Specifications

Density 1.0100g/mL
Boiling Point 108.0°C to 110.0°C (66.0 Mbar)
Flash Point 67°C
Refractive Index 1.5428 to 1.5448
Quantity 100 g
Specific Gravity 1.01
Formula Weight 119.17
Chemical Name or Material 2,3-Cyclopentenopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.