Learn More
Medchemexpress LLC 2,3-dehydro-2-deoxy-N-acetylneuraminic acid | 24967-27-9 | MFCD00135810 | 98.0%, ≥99.0% | 291.25 | C11H17NO8 | 1MG
SDP
Catalog No. 5000239100
Encompass_Preferred
2,3-Dehydro-2-deoxy-N-acetylneuraminic acid (Neu5Ac2en, DANA) is a neuraminidase (sialidase) inhibitor used in biochemical and antiviral research. It inhibits human neuraminidase isoforms NEU1-NEU4 (reported IC50s: NEU1 143 μM, NEU2 43 μM, NEU3 61 μM, NEU4 74 μM) and has demonstrated anti-influenza virus activity. The compound is supplied in multiple pack sizes and purity grades for research applications.
  • Pan-sialidase (neuraminidase) inhibitor.
  • Reported IC50s for NEU1-NEU4: 143 μM, 43 μM, 61 μM, 74 μM.
  • Demonstrated anti-influenza virus activity.
  • Molecular formula C11H17NO8 and molecular weight 291.25.
  • Available in multiple pack sizes and purity grades for research use.
Catalog No. 50-002-39100 Supplier Medchemexpress LLC Supplier No. HY1257981MG
2,3-Dehydro-2-deoxy-N-acetylneuraminic acid (Neu5Ac2en, DANA) is a neuraminidase (sialidase) inhibitor used in biochemical and antiviral research. It inhibits human neuraminidase isoforms NEU1-NEU4 (reported IC50s: NEU1 143 μM, NEU2 43 μM, NEU3 61 μM, NEU4 74 μM) and has demonstrated anti-influenza virus activity. The compound is supplied in multiple pack sizes and purity grades for research applications.
  • Pan-sialidase (neuraminidase) inhibitor.
  • Reported IC50s for NEU1-NEU4: 143 μM, 43 μM, 61 μM, 74 μM.
  • Demonstrated anti-influenza virus activity.
  • Molecular formula C11H17NO8 and molecular weight 291.25.
  • Available in multiple pack sizes and purity grades for research use.