Learn More
Medchemexpress LLC 2,3-Di(furan-2-yl)quinoxaline | 57490-73-0 | C16H10N2O2 | 1 ML
SDP
Catalog No. 5000372482
Encompass_Preferred
2,3-Di(furan-2-yl)quinoxaline is a quinoxaline that exhibits blue fluorescence. It is cell permeable and sufficiently bright at micromolar concentrations, making it suitable for research applications as a fluorescent dye.
  • Exhibits blue fluorescence
  • Cell permeable
  • Sufficiently bright at micromolar concentration
  • For research use only
Catalog No. 50-003-72482 Supplier Medchemexpress LLC Supplier No. HY15334910MM/1ML
2,3-Di(furan-2-yl)quinoxaline is a quinoxaline that exhibits blue fluorescence. It is cell permeable and sufficiently bright at micromolar concentrations, making it suitable for research applications as a fluorescent dye.
  • Exhibits blue fluorescence
  • Cell permeable
  • Sufficiently bright at micromolar concentration
  • For research use only