Learn More
Medchemexpress LLC 2,3-Di(furan-2-yl)quinoxaline | 57490-73-0 | C16H10N2O2 | 100 MG
SDP
Catalog No. 5000372527
Encompass_Preferred
2,3-Di(furan-2-yl)quinoxaline is a quinoxaline that exhibits blue fluorescence.
  • Cell permeable
  • Sufficiently bright at a micromolar concentration (1.5 μM)
Catalog No. 50-003-72527 Supplier Medchemexpress LLC Supplier No. HY153349100MG
2,3-Di(furan-2-yl)quinoxaline is a quinoxaline that exhibits blue fluorescence.
  • Cell permeable
  • Sufficiently bright at a micromolar concentration (1.5 μM)