Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine 98.0+%, TCI America™
SDP

Catalog No. D49695G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49695G 5 g
D49691G 1 g
D496925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D49695G Supplier TCI America Supplier No. D49695G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 161599-46-8
Molecular Formula C13H16FN3O6
Molecular Weight (g/mol) 329.284
MDL Number MFCD07369302
InChI Key NWJBWNIUGNXJGO-RPULLILYSA-N
PubChem CID 11809635
IUPAC Name [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate
SMILES CC1C(C(C(O1)N2C=C(C(=NC2=O)N)F)OC(=O)C)OC(=O)C

Specifications

Melting Point 189°C
Color White
Quantity 5 g
Formula Weight 329.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2′,3′-Di-O-acetyl-5′-deoxy-5-fluorocytidine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.