Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diamino-5-bromo-6-methylpyridine, 95%, Thermo Scientific™

Catalog No. AAH66847MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66847MD 250 mg
AAH6684703 1 g
AAH6684706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH66847MD Supplier Thermo Scientific Chemicals Supplier No. H66847MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59352-90-8
Molecular Formula C6H8BrN3
Molecular Weight (g/mol) 202.055
MDL Number MFCD00128868
InChI Key XFHAYQGHHKCVBA-UHFFFAOYSA-N
Synonym 5-bromo-6-methyl-2,3-pyridinediamine, 2,3-diamino-5-bromo-6-picoline, 2,3-pyridinediamine, 5-bromo-6-methyl, 2,3-diamino-5-bromo-6-methylpyridine, 5-bromo-2,3-diamino-6-methylpyridine, 5-bromo-6-methyl-pyridine-2,3-diamine, 2,3-diamino-5-bromo-6-methylpyridine 2,3-diamino-5-bromo-6-picoline, 5,6-diamino-3-bromo-, a-picoline, pubchem5382, 3-bromo-5,6-diamino-2-picoline
PubChem CID 101022
IUPAC Name 5-bromo-6-methylpyridine-2,3-diamine
SMILES CC1=C(C=C(C(=N1)N)N)Br

Specifications

Color Brown to Yellow
Quantity 250 mg
Beilstein 123081
Formula Weight 202.06
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2,3-Diamino-5-bromo-6-methylpyridine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 261-714-8
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.