Learn More
Medchemexpress LLC 2,3-Diaminonaphthalene | 771-97-1 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYD007310MM/1ML
Encompass_Preferred
2,3 Diaminonaphthalene is described as a highly selective colorimetric and fluorometric reagent. It is used for the detection of selenium ions and for the fluorometric determination of nitrite.
  • Highly selective colorimetric and fluorometric reagent for selenium ion detection.
  • Used for fluorometric determination of nitrite.
  • Available in various sizes, including solid and solution forms.
  • Provided with a data sheet, COA, SDS, and handling instructions.
  • Soluble in DMSO.
Catalog No. 50-004-10841