Learn More
Medchemexpress LLC 2,3-Diaminonaphthalene | 771-97-1 | 98.0% | 5 G
SDP
Supplier:  Medchemexpress LLC HYD00735G
Encompass_Preferred
2,3-Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium ion detection and is also used for the fluorometric determination of nitrite. It is intended for research use only.
  • Highly selective colorimetric reagent for selenium ion detection.
  • Highly selective fluorometric reagent for selenium ion detection.
  • Used for the fluorometric determination of nitrite.
  • Intended for research use only.
Catalog No. 50-004-10662