Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diaminonaphthalene, 97%

Catalog No. AAA1299306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1299306 5 g
AAA1299303 1 g
AAA1299314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1299306 Supplier Thermo Scientific Chemicals Supplier No. A1299306
  • $581.00
    $481.65 / Each of 1

    Save $99.35

    17% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

CAS: 771-97-1 | C10H10N2 | 158.204 g/mol

2,3 Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium detection and also used for the fluorometric determination of nitrite. It is also useful for detection of ketones and alpha-keto acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3 Diaminonaphthalene is a highly selective colorimetric and fluorometric reagent for selenium detection and also used for the fluorometric determination of nitrite. It is also useful for detection of ketones and alpha-keto acids.

Solubility
Slightly soluble in water. Soluble in pyridine, dimethylsulfoxide, dimethyformamide, dil.hydrochloric acid, ethanol, ether and hot methanol.

Notes
Incompatible with oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-97-1
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00004116
InChI Key XTBLDMQMUSHDEN-UHFFFAOYSA-N
Synonym 2,3-diaminonaphthalene, 2,3-naphthalenediamine, 2,3-naphthylenediamine, unii-2bnz6brs87, 2bnz6brs87, ccris 8399, zlchem 848, 2,3-diaminonapthalene, 2,3 diaminonapthalene, pubchem15532
PubChem CID 69872
IUPAC Name naphthalene-2,3-diamine
SMILES C1=CC=C2C=C(C(=CC2=C1)N)N

Specifications

Melting Point 193°C to 196°C
Density 1.096
Quantity 5 g
Beilstein 2206394
Solubility Information Slightly soluble in water; Soluble in pyridine,dimethylsulfoxide,dimethyformamide,dil;hydrochloric acid,ethanol,ether and hot methanol.
Formula Weight 158.2
Percent Purity 97%
Chemical Name or Material 2,3-Diaminonaphthalene
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H350-H302-H312-H332-H315-H319-H335
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 212-241-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.