Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diaminonaphthalene, Sigma-Aldrich

Catalog No. 01258999 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-999 5 g
01-258-930 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-999 Supplier MilliporeSigma Supelco Supplier No. D27575G
Add to Cart
Add to Cart

2,3-Diaminonaphthalene is also known as 2,3-Naphthalenediamine or DAN. It is one of the isomers of naphthalene bearing two amino groups, which are also called naphthalenediamines. They are usually prepared by reducing the nitronaphthalene derivative and aminating the subsequent hydroxynaphthalene derivative.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-97-1
Molecular Weight (g/mol) 158.2 g/mol

Specifications

Quantity 5 g
Percent Purity ≥95%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.