Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D4597200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4597200MG 200 mg
D45971G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4597200MG Supplier TCI America Supplier No. D4597200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 13214-70-5
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.97
MDL Number MFCD00021589
InChI Key GTILXPRQNNYDHT-UHFFFAOYSA-N
Synonym naphthalene, 2,3-dibromo, 2,3-dibromo-naphthalene, acmc-209uxy, pbn-10
PubChem CID 123298
IUPAC Name 2,3-dibromonaphthalene
SMILES BrC1=C(Br)C=C2C=CC=CC2=C1

Specifications

Melting Point 142°C
Color White-Yellow
Quantity 200 mg
Formula Weight 285.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.