Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropionamide, 97%, Thermo Scientific™

Catalog No. AAL1097806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1097806 5 g
AAL1097814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1097806 Supplier Thermo Scientific Chemicals Supplier No. L1097806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,3-Dibromopropionamide, is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15102-42-8
Molecular Formula C3H5Br2NO
Molecular Weight (g/mol) 230.887
MDL Number MFCD00031484
InChI Key DZQCMQRQFZXQKN-UHFFFAOYSA-N
PubChem CID 85798
IUPAC Name 2,3-dibromopropanamide
SMILES C(C(C(=O)N)Br)Br

Specifications

Melting Point 132°C to 133°C
Quantity 5 g
Beilstein 1749448
Synonym 2,3-dibromopropionamide, propanamide, 2,3-dibromo, acmc-209twc, propionamide,3-dibromo, 2,3-dibromo-propionamide, #, propanamide,2,3-dibromo, propionamide, 2,3-dibromo, ksc493i7n, 2,3-dibromopropionamide standardforacrylamideglcdetermination
Solubility Information Insoluble in water.
Formula Weight 230.9
Percent Purity 97%
Chemical Name or Material 2,3-Dibromopropionamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 239-153-5
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.