Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropionamide 98.0+%, TCI America™
SDP

Catalog No. D137825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D137825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D137825G Supplier TCI America Supplier No. D137825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15102-42-8
Molecular Formula C3H5Br2NO
Molecular Weight (g/mol) 230.887
MDL Number MFCD00031484
InChI Key DZQCMQRQFZXQKN-UHFFFAOYSA-N
Synonym 2,3-dibromopropionamide, propanamide, 2,3-dibromo, acmc-209twc, propionamide,3-dibromo, 2,3-dibromo-propionamide, #, propanamide,2,3-dibromo, propionamide, 2,3-dibromo, ksc493i7n, 2,3-dibromopropionamide standardforacrylamideglcdetermination
PubChem CID 85798
IUPAC Name 2,3-dibromopropanamide
SMILES C(C(C(=O)N)Br)Br

Specifications

Melting Point 133°C
Color White
Quantity 25 g
Formula Weight 230.89
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dibromopropionamide [Standard for Acrylamide GLC Determination]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.