Learn More
Crescent Chemical Co Inc 2,3-DIBROMOPROPIONIC ACID 1ML
SDP
Catalog No. 5069159
Encompass_Preferred
2,3-Dibromopropionic acid Single Component Calibration Standard in Methyl tert.-butyl ether CAS 600-05-5 Size - 1mL Catalog Number - 3461MT.10
Catalog No. 50-691-59 Supplier Crescent Chemical Co Inc Supplier No. CT3461MT.10
2,3-Dibromopropionic acid Single Component Calibration Standard in Methyl tert.-butyl ether CAS 600-05-5 Size - 1mL Catalog Number - 3461MT.10